Skip to Content
Merck
All Photos(1)

Key Documents

344206-M

Millipore

Formamide, Molecular Biology Grade

Formamide, Molecular Biology Grade

Deionized. Adenine deaminase inhibitor. Used in DNA stripping during genetic fingerprinting techniques.

Synonym(s):

Formamide, CH₃NO, Amide C1, Formic amide

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
391,00 kr.
1 L
2.700,00 kr.

391,00 kr.


In StockDetails


Request a Bulk Order

Select a Size

Change View
100 ML
391,00 kr.
1 L
2.700,00 kr.

About This Item

Linear Formula:
HCONH2
CAS Number:
Molecular Weight:
45.04
Beilstein:
505995
MDL number:
UNSPSC Code:
12352111
NACRES:
NA.28
grade:
Molecular Biology
technique(s):
hybridization: suitable
bp:
210 °C (lit.)
vapor pressure:
0.08 mmHg ( 20 °C)
30 mmHg ( 129 °C)

391,00 kr.


In StockDetails


Request a Bulk Order

grade

Molecular Biology

Quality Level

vapor density

1.55 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)
30 mmHg ( 129 °C)

form

liquid

autoignition temp.

932 °F

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

technique(s)

hybridization: suitable

refractive index

n20/D 1.447 (lit.)

pH

8-10 (20 °C, 200 g/L)

bp

210 °C (lit.)

mp

2-3 °C (lit.)

solubility

water: 20 mg/mL (~0.5 M)

density

1.134 g/mL at 25 °C (lit.)

cation traces

heavy metals (as Pb): ≤0.0001%

foreign activity

DNases, proteases, RNases, none detected

storage temp.

10-30°C

SMILES string

NC=O

InChI

1S/CH3NO/c2-1-3/h1H,(H2,2,3)

InChI key

ZHNUHDYFZUAESO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Deionized. Adenine deaminase inhibitor. Used in DNA stripping during genetic fingerprinting techniques.
Useful as an ionizing solvent. Absorbance (0.5 M): ≤0.08 at 260 nm; ≤0.03 at 280 nm. Adenine deaminase inhibitor. Used in DNA stripping during genetic fingerprinting techniques. Supplied as deionized.

Application

Formamide destabilizes nucleic acid duplexes and may be used, typically, at a concentration of 50%, in hybridization protocols requiring lower hybridization temperatures.

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

305.6 °F

Flash Point(C)

152 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service