Skip to Content
Merck
All Photos(1)

Key Documents

266785

Sigma-Aldrich

Diacylglycerol Kinase Inhibitor I

The Diacylglycerol Kinase Inhibitor I, also referenced under CAS 93076-89-2, controls the biological activity of Diacylglycerol Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Diacylglycerol Kinase Inhibitor I, 6-{2-{4-[(p-Fluorophenyl)phenylmethylene]-1-piperidinyl}ethyl}-7-methyl-5H-thiazolo(3,2-a)pyrimidine-5-one, R59022, R59022, 6-{2-{4-[(p-Fluorophenyl)phenylmethylene]-1-piperidinyl}ethyl}-7-methyl-5H-thiazolo(3,2-a)pyrimidine-5-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
1.280,00 kr.

1.280,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
5 MG
1.280,00 kr.

About This Item

Empirical Formula (Hill Notation):
C27H26FN3OS
CAS Number:
Molecular Weight:
459.58
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

1.280,00 kr.


Please contact Customer Service for Availability

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

ethanol: 12 mg/mL
DMSO: 4 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

Fc1ccc(cc1)C(=C3CCN(CC3)CCc4[c]([n]5c([s]cc5)nc4C)=O)c2ccccc2

InChI

1S/C27H26FN3OS/c1-19-24(26(32)31-17-18-33-27(31)29-19)13-16-30-14-11-22(12-15-30)25(20-5-3-2-4-6-20)21-7-9-23(28)10-8-21/h2-10,17-18H,11-16H2,1H3

InChI key

MFVJXLPANKSLLD-UHFFFAOYSA-N

General description

Potentiates the activity of protein kinase C by inhibiting diacylglycerol metabolism. Inhibits diacylglycerol kinase (IC50 = 2.8 µM) without affecting phosphodiesterase or phosphatidylinositol kinase. Potentiates this effect in studies of signal-transducing inositol lipid metabolism. Inhibits human platelet DAG kinase in intact cells (IC50 = 3.6 µM) and in isolated membranes (IC50 = 6.7 µM). In porcine thymus cytosol it inhibits the 80 kDa DAG kinase (IC50 = 10 µM) but has no effect on the 150 kDa DAG kinase. Weak dopamine D2 and histamine H1 antagonist and strong serotonin-S2 antagonist.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Diacylglycerol Kinase
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 3 months at 4°C.

Other Notes

Goin, M., et al. 1993. FEBS Lett. 316, 68.
Ohtsuka, T., et al. 1990. J. Biol. Chem.265, 15418.
de Chaffoy de Courcelles, D., et al. 1985. J. Biol. Chem.260, 15762.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service