Skip to Content
Merck
All Photos(1)

Key Documents

2500-S

Millipore

ReBlot Plus Western Blot Recycling

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
904,00 kr.

904,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 SET
904,00 kr.

About This Item

UNSPSC Code:
12352108
NACRES:
NB.23

904,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service