Skip to Content
Merck
All Photos(1)

Key Documents

234169

Sigma-Aldrich

Anti-Collagen Type I/III, Cyanogen Bromide Fragments Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

form

liquid

does not contain

preservative

species reactivity

human, rat, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−70°C

target post-translational modification

unmodified

Gene Information

human ... COL1A1(1277)

General description

Anti-Collagen Type I/III, Cyanogen Bromide Fragments, rabbit polyclonal specifically recognizes type I and III collagens. It is validated for WB, IF, IP, and IHC with frozen sections.
Rabbit polyclonal antibody supplied as undiluted serum. Recognizes type I and type III collagen.
Recognizes type I and type III collagens. Does not cross-react with fibronectin or type II, type V, or type VI collagen.

Immunogen

Fetal Mouse Skin
purified, fetal mouse skin collagens type I and III, digested with cyanogen bromide

Application

Frozen Sections (1:30-1:60, see comments)

Immunoblotting (1:200-1:500)

Immunofluorescence (1:20-1:40)

Immunoprecipitation (1:20-1:60)

Warning

Toxicity: Standard Handling (A)

Physical form

Undiluted serum.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Romanos, G.E., et al. 1992. J. Periodont. Res. 27, 101.
Zimmermann, B., et al. 1992. Eur. Arch. Biol. 103, 93.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luis Dos Santos et al.
International journal of cardiology. Heart & vasculature, 21, 22-28 (2018-09-28)
Atrial fibrillation (AF) frequently coexists with congestive heart failure (CHF). The increased susceptibility to AF in CHF has been attributed to a variety of structural and electrophysiological changes in the atria, particularly dilation and interstitial fibrosis. We evaluated atrial remodeling
Jennifer N Walker et al.
Plastic and reconstructive surgery. Global open, 7(2), e2037-e2037 (2019-03-19)
Bacterial contamination of breast implants causes infection, can lead to capsular contracture, and is implicated in breast implant-associated anaplastic large cell lymphoma. Bacteria, however, also colonize clinically benign breast implants and little is known about the biologic signals that trigger

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service