Skip to Content
Merck
All Photos(1)

Documents

23-047-M

Sigma-Aldrich

UbcH7 Protein, active, 10 µg

Active, full length, recombinant human UbcH7. N-terminal HA, 6His-tagged. For use in Enzyme Assays. Functions as an E2 enzyme in ubiquitination assays.

Synonym(s):

Ubiquitin-conjugating enzyme E2 L3, L-UBC, Ubiquitin carrier protein L3, Ubiquitin-conjugating enzyme E2-F1, Ubiquitin-protein ligase L3, UBE2L3, UBCE7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

biological source

human

Quality Level

recombinant

expressed in E. coli

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

UniProt accession no.

General description

Ubiquitin-conjugating (E2) enzymes are characterized by the presence of a highly conserved ubiquitin-conjugating domain, which accommodates ATP-activated ubiquitin (Ub) via a covalently linked thioester on its active-site residue. E2 enzymes act through selective protein-protein interactions with the ubiquitin-activating E1 enzyme and ubiquitin ligase E3 enzymes and are able to differentiate effects on downstream substrates, either with a single Ub molecule or a Ub chain. While E3s are involved in substrate selection, E2s are the main determinants for selection of the lysine to construct Ub chains, which thereby directly control the cellular fate of the substrate.

UbcH7 shows broad specificity for HECT-type E3s but does not function with most RING-containing E3 ubiquitin-protein ligases because it lacks intrinsic E3-independent reactivity with lysine. Despite lacking this lysine reactivity, UbcH7 exhibits activity with the RBR family E3 enzymes, such as Parkin and ARIH1.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service