Skip to Content
Merck
All Photos(2)

Documents

23-028-M

Sigma-Aldrich

IkBa Protein, activated, 10 µg

Activated, full length, recombinant human IκBα. N-terminal c-Myc, 6His-tagged. Phosphorylated in vitro using IKKα, active. For use in Enzyme Assays. Behaves as a substrate in ubiquitin assays.

Synonym(s):

NF-kappa-B inhibitor alpha, I-kappa-B-alpha, Major histocompatibility complex enhancer-binding protein MAD3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

biological source

human

Quality Level

100
300

recombinant

expressed in E. coli

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

UniProt accession no.

General description

IκBα is a cellular protein that inhibits the transcription factor NF-κB. NF-κB plays a central role in cellular stress and inflammatory responses by controlling cytokine-inducible gene expression and lymphocyte stimulation by antigens. In non-stimulated cells NF-κB is sequestered in the cytoplasm via the inhibitory protein IκBα, which obscures the nuclear localization signal of NF-κB and also blocks its ability to bind DNA. In response to various stimuli, including viral infection, ultraviolet radiation and inflammatory cytokines, IκBα is phosphorylated by the protein kinase complex IKK, which triggers its rapid degradation by ubiquitin mediated proteolysis. The liberated NF-κB then translocates to the nucleus and activates the expression of target genes. Mutations in the gene for IκBα result in the functional impairment of NF-κB which causes a decreased production of proinflammatory cytokines and certain interferons, rendering patients susceptible to infection.
N-terminal GST-tagged, recombinant human Smurf1 full length, expressed in E coli.

Application

Ubiquitination Cascade Comoponent: Substrate

Physical form

Purified using immobilized metal affinity chromatography.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service