Skip to Content
Merck
All Photos(1)

Key Documents

219399

Sigma-Aldrich

Cathepsin D Substrate I

Useful as a substrate for the determination of cathepsin D activity.

Synonym(s):

Cathepsin D Substrate I, Bz-RGFFP-4-MeOβNA, HCl

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
802,00 kr.

802,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
802,00 kr.

About This Item

Empirical Formula (Hill Notation):
C49H55N9O7 · xHCl
Molecular Weight:
882.02 (free base basis)
UNSPSC Code:
51111800
NACRES:
NA.77

802,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

white

solubility

DMSO: 5 mg/mL

fluorescence

λex ~345 nm
λem ~425 nm

shipped in

ambient

storage temp.

−20°C

General description

A useful substrate for colorimetric or fluorometric assay of cathepsin D activity. Cathepsin D hydrolyzes the Phe-Phe bond. Dipeptidyl aminopeptidase II is required to free the 4-methoxy-β-naphthylamine (4MβNA). Excitation max: ~345 nm; emission max: ~425 nm.
Useful as a substrate for the determination of cathepsin D activity. Suitable for either colorimetric or fluorometric assay. Cathepsin D hydrolyzes the Phe-Phe bond. Requires dipeptidyl aminopeptidase II to free 4-methoxy-β-naphthylamine (4MeOβNA).

Biochem/physiol Actions

Cell permeable: no
Primary Target
Substrate for the determination of cathepsin D activity
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

Bz-Arg-Gly-Phe-Phe-Pro-4MeOβNA, HCl

Other Notes

Takahashi, T., and Tang, J. 1981. Methods Enzymol.80, 565.
Smith, R.E., and Van Frank, R.M. 1975. in Lysosomes in Biology and Pathology4 (Dingle, J.T., and Dean, R.T., eds.) p. 193, American Elsevier, Holland.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service