Skip to Content
Merck
All Photos(1)

Documents

208773

Sigma-Aldrich

Calpain Substrate IV, Cell-Permeable, Fluorogenic

The non-permeant fluorogenic Calpain Substrate III is made cell-permeable by a c-terminal heptaarginine extension via a Glu linkage.

Synonym(s):

Calpain Substrate IV, Cell-Permeable, Fluorogenic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C127H206N50O27S
Molecular Weight:
2897.38
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

solubility

DMSO: 5 mg/mL

fluorescence

λex 480 nm; λem 320 nm

shipped in

ambient

storage temp.

−20°C

General description

The non-permeant fluorogenic Calpain Substrate III (Cat. No. 208771) is made cell-permeable by a c-terminal heptaarginine extension via a Glu linkage. In addition to the expected improvement in permeability, the modified peptide also exhibits enhanced Calpain B sensitivity (kcat = 0.2 s-1, kcat/KM = 5000 M-1 s-1) when compared to the parental peptide (kcat = 0.17 s-1, kcat/KM = 680 M-1 s-1). Its suitability in detecting cellular Ca2+-dependent calpain activity is demonstrated both in intact Drosophila melanogaster S2 cells following ionomycin stimulation and in S2 lysates via direct Ca2+ addition.
The non-permeant fluorogenic Calpain Substrate III (Cat. No. 208771) is made cell-permeable by a c-terminal heptaarginine extension via a Glu linkage. In addition to the expected improvement in permeability, the modified peptide also exhibits enhanced Calpain B sensitivity (kcat = 0.2 s-1, kcat/KM = 5000 M-1 s-1) when compared to the parental peptide (kcat = 0.17 s-1, kcat/KM = 680 M-1 s-1). Its suitability in detecting cellular Ca2+-dependent calpain activity is demonstrated both in intact Drosophila melanogaster S2 cells following ionomycin stimulation and in S2 lysates via direct Ca2+ addition.
The non-permeant fluorogenic Calpain Substrate III is made cell-permeable by a c-terminal heptaarginine extension via a Glu linkage.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

(DABCYL)-Thr-Pro-Leu-Lys~Ser-Pro-Pro-Pro-Ser-Pro-Arg-Glu(EDANS)-Arg-Arg-Arg-Arg-Arg-Arg-Arg-NH₂

Physical form

Supplied as a trifluoroacetate salt

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Banoczi, Z., et al. 2008. Bioconjug. Chem.19, 1375.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service