Skip to Content
Merck
All Photos(1)

Key Documents

1.94524

Supelco

2-Propanol

EMPLURA®

Synonym(s):

2-Propanol, sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Beilstein:
635639
MDL number:
UNSPSC Code:
12352104
bp:
82 °C (lit.)
vapor pressure:
33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

vapor density

2.1 (vs air)

vapor pressure

33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

product line

EMPLURA®

autoignition temp.

750 °F

expl. lim.

2.0-12.7 %, 93 °C

dilution

(low-cost alternative, for basic applications)

refractive index

n20/D 1.377 (lit.)

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

density

0.785 g/mL at 25 °C (lit.)

storage temp.

room temp

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

1.94524.0521: 500ML, Minimum order quantity: 20
1.94524.2521: 2.5L, Minimum order quantity: 4
1.94524.9027: 25 L Minimum Order Quantity: 1

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service