Skip to Content
Merck
All Photos(1)

Key Documents

1.93311

Supelco

Potassium sorbate

for analysis EMPARTA®

Synonym(s):

Potassium sorbate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7KO2
CAS Number:
Molecular Weight:
150.22
MDL number:
UNSPSC Code:
12352107
grade:
Laboratory Reagent
form:
solid
solubility:
water: soluble 1.95 g/L at at 20 °C
Pricing and availability is not currently available.

grade

Laboratory Reagent

vapor pressure

<0.01 hPa ( at 20 °C)

form

solid

pH

7.75-7.77 ( at 20.1 °C, 68.2 °F)

mp

(Decomposes before melting)

solubility

water: soluble 1.95 g/L at at 20 °C

storage temp.

room temp

SMILES string

[K+].[O-]C(=O)\C=C\C=C\C

InChI

1S/C6H8O2.K/c1-2-3-4-5-6(7)8;/h2-5H,1H3,(H,7,8);/q;+1/p-1/b3-2+,5-4+;

InChI key

CHHHXKFHOYLYRE-STWYSWDKSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

1.93311.1021: 1 kg, Minimum order quantity: 12
1.93311.5021: 5 kg, Minimum order quantity: 4

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service