Skip to Content
Merck
All Photos(1)

Key Documents

178494

Sigma-Aldrich

Apoptosis Inhibitor II, NS3694

The Apoptosis Inhibitor II, NS3694, also referenced under CAS 426834-38-0, controls the biological activity of Apoptosis. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Apoptosis Inhibitor II, NS3694, NS3694

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
1.270,00 kr.

1.270,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
10 MG
1.270,00 kr.

About This Item

Empirical Formula (Hill Notation):
C15H10ClF3N2O3
CAS Number:
Molecular Weight:
358.70
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

1.270,00 kr.


Please contact Customer Service for Availability

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 10 mg/mL
ethanol: slightly soluble

shipped in

ambient

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc(ccc1)NC(=O)Nc2c(ccc(c2)Cl)C(=O)O

InChI

1S/C15H10ClF3N2O3/c16-9-4-5-11(13(22)23)12(7-9)21-14(24)20-10-3-1-2-8(6-10)15(17,18)19/h1-7H,(H,22,23)(H2,20,21,24)

InChI key

GNCZTZCPXFDPLI-UHFFFAOYSA-N

General description

A cell-permeable diarylurea compound that specifically prevents the formation of the active ~700 kDa apoptosome complex induced by cytochrome c release. Alters cytochrome c and ATP-triggered association between Apaf-1 and caspase-9. Protects against apoptosome-mediated caspase activation and cell death (50 µM completely blocks TNF-α-induced death in MCF-casp3 cells). Unlike M50054 (Cat. No. 178488), NS3694 exhibits no effect on apoptosome-independent caspase activation and cell death induced by Fas ligand (FasL). Does not inhibit cytochrome c release or the activity of caspase-3 and -9.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Apoptosome complex formation
Product does not compete with ATP.
Reversible: no
Target IC50: 50 µM completely blocks TNF-α-induced death in MCF-casp3 cells

Warning

Toxicity: Standard Handling (A)

Other Notes

Lademann, U., et al. 2003. Mol. Cell. Biol.23, 7829.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service