Skip to Content
Merck
All Photos(1)

Key Documents

175580

Sigma-Aldrich

Angiogenesis Inhibitor

The Angiogenesis Inhibitor, also referenced under CAS 186611-44-9, controls the biological activity of Angiogenesis. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Angiogenesis Inhibitor, ( Z,E)-3-(Imidazol-4-ylmethylene)indolin-2-one, (Z,E)-3-(Imidazol-4-ylmethylene)indolin-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
1.280,00 kr.

1.280,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
10 MG
1.280,00 kr.

About This Item

Empirical Formula (Hill Notation):
C12H9N3O
CAS Number:
Molecular Weight:
211.22
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

1.280,00 kr.


Please contact Customer Service for Availability

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable indolinone compound that displays anti-angiogenesis properties (30% inhibition of control at 10 µM in an in vitro rat aortic ring model ) with a potency that is comparable to that of SU5416 (Cat. No. 676487; 22% inhibition of control at 10 µM). Acts as a moderate ATP-competitive inhibitor of hEGF-R tyrosine kinase activity (54% inhibition at 10 µM).
A cell-permeable indolinone compound that displays anti-angiogenesis properties (30% inhibition of control at 10 µM in an in vitro rat aortic ring model). The potency has been reported to be comparable to that of SU5416 (Cat. No. 676487; 22% inhibition of control at 10 µM). Acts as a moderate ATP-competitive inhibitor of hEGF-R tyrosine kinase activity (54% inhibition at 10 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
hEGF-R tyrosine kinase activity
Product competes with ATP.
Reversible: no
Target IC50: 54% inhibition at 10 µM against hEGF-R tyrosine kinase activity

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Other Notes

Braud, E., et al. 2003. J. Enzyme Inhib. Med. Chem.18, 243.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service