Skip to Content
Merck
All Photos(1)

Documents

17-10523

Sigma-Aldrich

EZ-Magna NuCLEAR RIP (Native) Nuclear RNA-Binding Protein Immunoprecipitation Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105331
NACRES:
NA.84

Quality Level

General description

Magna Nuclear RIP Kits are specially designed to allow the discovery and analysis of a variety of chromatin associated RNAs such as long non-coding RNAs, enhancer RNAs and miRNAs . These chromatin-associated RNAs often regulate gene expression and can be analyzed with applications including quantitative reverse transcription polymerase chain reaction (RT-PCR), microarray analysis (RIP-chip) and next generation sequencing (RIP-Seq).

Nuclear RIP can be performed using chromatin that has interactions stabilized by formaldehyde treatment (cross-linked) or chromatin that has not been treated with a cross linking reagent (native). While both of these approaches are similar in that they are designed to recover chromatin associated RNA, the reagents used and the details of the protocol and types of interactions typically detected are different. Native RIP is expected to recover high affinity, more direct interactions between proteins encoded RNA binding motifs and candidate RNAs. In contrast, nuclear RIP with cross-linked chromatin can capture higher molecular weight complexes in in vivo configurations with possibly lower affinities. For less well understood proteins and protein complexes often both approaches are used.

Application

EZ-Magna Nuclear RIP (Native) RNA-Binding Protein Immunoprecipitation Kit is designed for the analysis of chromatin associated RNA such lncRNAs, enhancer RNAs and miRNAs.

Packaging

Kit capacity: 24 RNA-binding protein immunoprecipitation assays using a native (non-cross-linked) nuclear lysate

Components

Nuclei Isolation Buffer ;RIP Lysis Buffer;Protein A/G Magnetic Beads;RIP Lysis Buffer;0.5M EDTA;DNase I (RNase Free) 2 U/μL;DNase I Reaction Buffer ;Protease Inhibitor Cocktail III, Animal Free;RNAse Inhibitor;Control Antibodies and PrimersNormal Mouse IgG Negative Control Antibody;Anti-EZH2 Positive Control Antibody ;NEAT1 Positive Control Primers ;U1 snRNA Negative Control Primers

Legal Information

NuCLEAR is a trademark of Sigma-Aldrich Co. LLC

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service