Skip to Content
Merck
All Photos(1)

Key Documents

14-783-M

Sigma-Aldrich

PI3 Kinase (p110α(E545K)/p85α), 10 µg

Complex of N-terminal 6His-tagged, recombinant, full-length human p110α containing the mutation E545K & untagged, recombinant, full-length human p85α. For use in Kinase Assays.

Synonym(s):

PI 3-K, Phosphatidylinositol 3-kinase complex class IA, p110alpha/p85alpha, p110-alpha, phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha, phosphoinositide-3-kinase

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
4.350,00 kr.
250 μG
27.960,00 kr.

4.350,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
10 μG
4.350,00 kr.
250 μG
27.960,00 kr.

About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.32

4.350,00 kr.


Please contact Customer Service for Availability

biological source

human

Quality Level

recombinant

expressed in Sf21 cells

form

liquid

mol wt

Mw 125.3 kDa (p110α; E545K)
Mw 83.7 kDa (p85α)

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

color

colorless

solubility

water: soluble

NCBI accession no.

UniProt accession no.

storage temp.

−70°C

Gene Information

human ... PIK3CA(5290)

General description

Research area: Cell signaling

PI3 Kinase is a complex of N-terminal 6His-tagged, recombinant, full-length human p110α containing the mutation E545K and untagged, recombinant, full-length human p85α.

Biochem/physiol Actions

p110α (PIK3CA)/p85α (PIK3R1) isoform of phosphoinositide 3-kinase (PI3K), plays a critical role in the regulation of various cellular processes. These include cellular survival, proliferation, migration, invasion, and metastasis.[1] Additionally, the signaling pathway involving the p110α isoform of PI3K is responsible for triggering physiological cardiac hypertrophy induced by exercise.[2] PIK3 also plays a vital role in activating Protein kinase B or Akt (PKB/Akt), an essential component of insulin and growth factors signaling pathways. Furthermore, PIK3 participates in synthesizing PIP3 (phosphatidylinositol (3,4,5)-trisphosphate), a key signaling molecule within the PI3K/Akt pathway. These significant functions highlight the critical role of PIK3 in mediating important cellular signaling processes, including inositol phosphate metabolism.[3] PIK3CA mutations can alter the tumor immunogenicity, which potentially affects its susceptibility to immunotherapies. The E545K substitution is a somatic mutation in p110α that has been associated with tumors of the colon and brain. Combined in vitro and in vivo studies have shown that this mutation confers higher lipid kinase activity than the wild type and is able to induce oncogenic transformation.[4]

Packaging

Also available in 250 μg size --call for pricing and availability and reference catalog number 14-783M when ordering the 250 μg size.

Storage and Stability

Store at -70°C from date of shipment. For maximum recovery of product, centrifuge original vial prior to removing the cap.

Rapidly thaw the vial under cold water and immediately place on ice. Aliquot unused material into pre-chilled microcentrifuge tubes and immediately snap-freeze the vials in liquid nitrogen prior to re-storage at -70°C.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service