Skip to Content
Merck
All Photos(1)

Documents

1.46820

Millipore

Tryptic Soy Agar - Ready-to-use Settle Plates

Sodium thiosulfate, Histidine, Tween®, Lecithin, ICR plus lockable plate, sterile; γ-irradiated, plate diam. 150 mm, pkg of Triple packed, suitable for air monitoring

Synonym(s):

TSA, Soybean Casein Digest Agar, ICR settle plate,, Tryptic Soy Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

sterile; γ-irradiated

form

plate

feature

2D bar code
ready-to-use

composition

Histidine
Lecithin
Sodium thiosulfate
Tween®

packaging

pkg of (18 plates per box)
pkg of Triple packed

plate diam.

150 mm

pH

(7.3 (in H2O))

application(s)

air monitoring
microbiology
pharmaceutical

storage temp.

15-25°C

suitability

bacteria
fungi

Looking for similar products? Visit Product Comparison Guide

General description

Tryptic Soy Agar + LTHThio – ICR settle plate is designed for the determination of the total microbial aerobic count of microorganisms in isolators and clean rooms. Settle plates are used for air and personnel monitoring. Tryptic soy agar is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The formulation of the Soybean Casein-Digest medium contains a combination of casein and soy peptones supplying organic nitrogen, particularly amino acids and longer-chained peptides. The medium is supplemented with Lecithin, polysorbate 80 (Tween®), Histidine, Sodium thiosulfate and pyruvate to inactivate potential residues of disinfectants on dry surfaces. The plates are triple packed and gamma-irradiated in the final packaging.

Application

Tryptic Soy Agar (TSA, Soybean Casein Digest Agar) is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The media is used for air monitoring as well to monitor personnel (fingerprints).

Packaging

1468200018: 18 Plates per box

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service