Skip to Content
Merck
All Photos(1)

Key Documents

14-678-M

Sigma-Aldrich

WNK2 Protein, active, 10 µg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf21 cells

Quality Level

description

protein target WNK2

purified by

(Ni2+/NTA-agarose)

technique(s)

activity assay: suitable

UniProt accession no.

Gene Information

human ... WNK2(65268)

General description

N-terminal 6His-tagged, recombinant, human WNK2, amino acids 166-489

Quality

routinely evaluated by phosphorylation of myelin basic protein (Catalog # 13-104)

Physical properties

41.2kDa

Storage and Stability

Stable for 6 months at -70°C from date of shipment

Analysis Note

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Other Notes

COFACTOR: Magnesium (By similarity).
SIZE: 2297 amino acids; 242676 Da
TISSUE SPECIFICITY: Predominantly expressed in heart, brain and colon.
SIMILARITY: SwissProt: Q9Y3S1 ## Belongs to the protein kinase superfamily. Ser/Thr protein kinase family. WNK subfamily. & Contains 1 protein kinase domain.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service