Skip to Content
Merck
All Photos(1)

Key Documents

1.46690

Millipore

Minimal Glucose Agar Settle Plate

90 mm settle plates are designed for AMES test to examine chemical components for toxic/cancerogenic properties.

Synonym(s):

Ames Agar

Sign Into View Organizational & Contract Pricing

Select a Size

120 PKG
2.080,00 kr.

2.080,00 kr.


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
120 PKG
2.080,00 kr.

About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

2.080,00 kr.


Please contact Customer Service for Availability

Request more information

Quality Level

sterility

sterile

form

plate

feature

ready-to-use
ICR Settle plate

packaging

box of 120 plates

technique(s)

microbiological culture: suitable

plate diam. × filling volume

90 mm × 20 mL

pH

7 ( in H2O)

application(s)

bioburden testing
industrial qc
life science and biopharma
microbiology
pharmaceutical

storage temp.

15-25°C

General description

Minimal Glucose Agar is used for the microbial mutagenicity Ames test. It is a bacterial bioassay performed in vitro to evaluate the mutagenicity of various environmental carcinogens and toxins. The principle is to detect point mutations using amino acid requiring strains. The Minimal Glucose Agar plate is taken as a bottom agar for the mutagenicity assay and includes the minimum ingredients for bacterial growth. Phosphates act as the buffering system. Glucose and Citrate are the carbon sources to produce energy. Magnesium is a cofactor for many metabolic reactions

Application

Minimal Glucose Agar for Ames test is used to identify the revert mutations which are present in strains. It can also be used to detect the mutagenicity of environmental samples such as drugs, dyes, reagents, cosmetics, wastewater, pesticides, and other substances which are easily solubilized in a liquid suspension.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service