Skip to Content
Merck
All Photos(1)

Key Documents

1.46559

Millipore

Columbia Blood Agar Settle plate

90 mm settle plate for the isolation and cultivation of fastidious bacteria as well as for detection of hemolysis.

Synonym(s):

Columbia Agar, Columbia Blood Agar Pharm.

Sign Into View Organizational & Contract Pricing

Select a Size

20 PKG
369,00 kr.

369,00 kr.


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
20 PKG
369,00 kr.

About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

369,00 kr.


Please contact Customer Service for Availability

Request more information

Quality Level

sterility

sterile

form

plate

feature

ready-to-use

packaging

pkg of (20 or 120 plates per box)

technique(s)

microbiological culture: suitable

plate diam. × volume

90 mm × 19 mL

pH

7.3 ( in H2O)

application(s)

microbiology
pharmaceutical

storage temp.

2-8°C

suitability

nonselective for

General description

Columbia Blood Agar settle plates supports rapid and luxuriant growth of fastidious and non-fastidious organisms. Also, this medium promotes typical colonial morphology, better pigment production, and more sharply defined hemolytic reactions. Starch serves as an energy source and neutralizes toxic metabolites. Sheep blood permits the detection of hemolysis and provides heme (X factor), which is required for the growth of many bacteria. Columbia Blood Agar is also appropriate for the cultivation of anaerobic bacteria or yeasts.

Application

Columbia Blood Agar is suitable for the isolation and cultivation of fastidious and non-fastidious organisms as well as for detection of hemolysis in various clinical and Pharmaceutical samples.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service