Skip to Content
Merck
All Photos(1)

Key Documents

14-645-M

Sigma-Aldrich

IRR Protein, active, 10 µg

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
3.450,00 kr.
250 μG
26.850,00 kr.

3.450,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
3.450,00 kr.
250 μG
26.850,00 kr.

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

3.450,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

description

protein target IRR

purified by

(Ni2+/NTA-agarose)

technique(s)

activity assay: suitable

UniProt accession no.

Gene Information

human ... INSRR(3645)

General description

FUNCTION: SwissProt: P14616 # This receptor probably binds an insulin related protein and has a tyrosine-protein kinase activity. It phosphorylates the insulin receptor substrates IRS-1 and IRS-2.
SIZE: 1297 amino acids; 143720 Da
SUBUNIT: Probable tetramer of 2 alpha and 2 beta chains linked by disulfide bonds. The alpha chains contribute to the formation of the ligand-binding domain, while the beta chains carry the kinase domain.
SUBCELLULAR LOCATION: Membrane; Single-pass type I membrane protein.
SIMILARITY: SwissProt: P14616 ## Belongs to the protein kinase superfamily. Tyr protein kinase family. Insulin receptor subfamily. & Contains 4 fibronectin type-III domains. & Contains 1 protein kinase domain.

Packaging

Also available in 10 μg size (cat#14-645) and in bulk (cat#14-645-K).

Quality

Routinely evaluated by phosphorylation of MBP (13-104).
routinely evaluated by phosphorylation of MBP (13-104).

Physical properties

37.9kDa

Storage and Stability

6 months at -70°C

Analysis Note

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service