Skip to Content
Merck
All Photos(1)

Documents

14-520-M

Sigma-Aldrich

Casein Kinase 1δ (aa 1-294) Protein, active, 10 µg

Active, N-terminal, GST-tagged, recombinant human CK1 delta, amino acids 1-294, for use in Kinase Assays.

Synonym(s):

Kinase protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in E. coli

mol wt

Mw 61 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

Gene Information

human ... CSNK1D(1453)

General description

Research area: Cell signaling

Research Category: Metabolism Research

Subcategory: Obesity

Protein Target: CK1γδ

Target Sub-Family: CK1

Casein Kinase 1δ (CK1δ ) is a serine/threonine kinase that belong to the family of casein kinase 1. The protein structure consists of anti-parallel β-sheet strands at N-terminal lobe (N-lobe) and α-helices and loop structures at the larger C-terminal lobe (C-lobe).

Application

Research Category
Metabolism
Research Sub Category
Obesity

Metabolic Disorders

Biochem/physiol Actions

Casein Kinase 1δ (CK1δ) is implicated in multiple cellular processes, including membrane trafficking, cytokinesis, circadian rhythm, and tumorigenesis. It interacts with critical molecules within cancer-related signaling pathways, such as Wnt/β-catenin, PI3K/AKT, and Hedgehog. Therefore, mutations or overexpression of the CK1δ coding gene can potentially contribute to the onset and progression of cancer.

Packaging

Also available in 250 µg size --call for pricing and availability and reference catalog number 14-520M when ordering the 250 µg size.

Quality

routinely evaluated by phosphorylation of KRRRALS(p)VASLPGL

Physical form

Glutathione-agarose

Storage and Stability

6 months at -20°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure, regulation, and (patho-)physiological functions of the stress-induced protein kinase CK1 delta (CSNK1D)
Xu P, et al.
Gene: X, 2019-2019 (2019)
Structure, regulation, and (patho-)physiological functions of the stress-induced protein kinase CK1 delta (CSNK1D)
Xu P, et al.
A Manual for Genetic Engineering: Advanced Bacterial Genetics, 715 (2019)
Francesca Vena et al.
Molecular cancer therapeutics, 19(8), 1623-1635 (2020-05-21)
Although gemcitabine is the cornerstone of care for pancreatic ductal adenocarcinoma (PDA), patients lack durable responses and relapse is inevitable. While the underlying mechanisms leading to gemcitabine resistance are likely to be multifactorial, there is a strong association between activating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service