Skip to Content
Merck

1.37031

SAFC

Sodium hydroxide solution

1 M (mol/L), EMPROVE® EXPERT

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352305

vapor pressure

3 mmHg ( 37 °C)

Quality Level

product line

EMPROVE® EXPERT

form

liquid

concentration

1 M (mol/L)

impurities

≤10 ppm Al
≤3 ppm Fe
≤30 ppm Sulfate
≤5 ppm Chloride
≤5 ppm Pd

storage temp.

no temp limit

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Filtered with 0.2 μm

Application

This product has been manufactured from Sodium hydroxide pellets Ph Eur, BP, JP, NF and water for injection USP. Water for injection USP has been prepared for this product by using purified water according to Ph Eur, JP.
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service