Skip to Content
Merck
All Photos(1)

Key Documents

12-547

Sigma-Aldrich

JAK3tide

JAK3tide primarily used in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

1000 μG
2.340,00 kr.

2.340,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
1000 μG
2.340,00 kr.

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

2.340,00 kr.


Please contact Customer Service for Availability

Quality Level

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Quality

Routinely evaluated as a substrate using JAK3, active (14-629)

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service