Skip to Content
Merck
All Photos(1)

Key Documents

1.25256

Supelco

Chromolith® Prep end cap set

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115716

product line

Chromolith®

Quality Level

technique(s)

HPLC: suitable

storage temp.

no temp limit

Analysis Note

visual control: conforms

Legal Information

Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service