Documents
1.17364
Millipore
MAS-100 Atmos® Silicone Gasket 34/10 mm
for use with MAS-100 Atmos®, autoclavable
About This Item
UNSPSC Code:
41103403
NACRES:
NA.74
material
silicone gasket
feature
autoclavable
manufacturer/tradename
MAS-100 Atmos®
application(s)
compressed gas monitoring
cosmetics
food and beverages
pharmaceutical
compatibility
for use with MAS-100 Atmos®
General description
MAS-100 Atmos® Silicone Gasket 34/10 mm is an accessory designed to secure pressure tube connection to the MAS-100 Atmos® microbial compressed gas sampler. This will avoid any potential leakage during sampling process.
Place the silicone gasket (ref. 1.17364) on top of the closed sampling head and fix the pressure connection tubing to the sampling head with the 34 mm tri-clamp (ref.1.17367).
Application
MAS-100 Atmos® Silicone Gasket 34/10 mm is used to tighten and secure the pressure tube connection to the sampler head.
Legal Information
MAS-100 Atmos is a registered trademark of MBV AG