Skip to Content
Merck
All Photos(1)

Documents

1.17057

Supelco

Spectromelt® A50

lithium metaborate 50 %, lithium tetraborate 50 %

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105
NACRES:
NA.21

composition

lithium metaborate, 50%
lithium tetraborate, 50%

Quality Level

storage temp.

2-30°C

General description

50% lithium tetraborate / 50% lithium metaborate

Legal Information

SPECTROMELT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service