Skip to Content
Merck
All Photos(1)

Key Documents

1.16730

Supelco

Blank Test Strips, reflectometric

REFLECTOQUANT®, for use with RQflex® 20

Sign Into View Organizational & Contract Pricing

Select a Size

50 TESTS
293,00 kr.

293,00 kr.


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
50 TESTS
293,00 kr.

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

293,00 kr.


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Product Name

Blank Strips, for use with REFLECTOQUANT®

description

quantitative

analyte chemical class(es)

sugars

specific analyte(s)

ammonium, ascorbic acid, calcium, chlorine, formaldehyde, hydrogen peroxide, hydroxymethylfurfural (Hmf), iron, lactic acid, magnesium, magnesium, malic acid, nitrate, nitrite, peracetic acid, phosphate, potassium, sulfite

compatibility

RQflex® 20
for use with REFLECTOQUANT®

detection method

reflectometric

storage temp.

15-25°C

General description

The Reflectoquant® Blank test strips incorporate paperzones not impregnated with reagent. They are used to perform tests to see whether the color of a sample (e.g. red juice) stays on the paper of the blank zones. A difference in color can indicate that the intrinsic color of the sample may affect results obtained with other Reflectoquant® test strips. For decolorization we then suggest to pre-treat the sample with Polyvinylpolypyrrolidone (Divergan®, 1.07302). Various application notes for these special samples are available.

Analysis Note

Application test: conforms

Legal Information

Divergan is a registered trademark of BASF SE
REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Hydroxymethylfurfural (HMF) in honey – reflectometric determination after after conversion with a barbituric acid derivative and an aminophenazone derivative

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service