Skip to Content
Merck
All Photos(1)

Key Documents

1.10537

Supelco

Carrez Clarification Kit

suitable for sample preparation, input: food(s), 5 ×

Sign Into View Organizational & Contract Pricing

Select a Size

2 X 100 ML
1.120,00 kr.

1.120,00 kr.


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
2 X 100 ML
1.120,00 kr.

About This Item

UNSPSC Code:
12352402
NACRES:
NA.21

1.120,00 kr.


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Quality Level

concentration

5 ×

technique(s)

photometry: suitable

input

food(s)

application(s)

sample preparation

storage temp.

no temp limit

General description

reagent kit for sample preparation in food analysis, 5-fold concentrate

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Quantitative analysis of Hydroxymethylfurfural in honey using a Chromolith® HighResolution RP-18e column without the common Carrez clarification step.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service