Skip to Content
Merck
All Photos(1)

Documents

1.09225

Millipore

MAS-100® case

For safety carrying of MAS-100®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103403

packaging

pkg of 1 kit

application(s)

cosmetics
food and beverages
pharmaceutical

compatibility

for use with MAS-100 NT® Ex
for use with MAS-100 NT®
for use with MAS-100®

storage temp.

room temp

General description

MAS-100® case is an additional accessory for the MAS-100® air sampler. The MAS-100® air sampler is designed to monitor air microbiology in cleanrooms and hygienically ultra-sensitive areas. Its high recovery rate is based on the Andersen impaction principle. An integrated airflow sensor compensates for differences in pressure and temperature in the environment. This practical transport case protects the device from damage and dirt and offers space to hold instructions and certificates as well as power adapter, connecting cable and dust cover.

Application

MAS-100® case is an additional accessory for the MAS-100® Air Sampler that protects the device from damage and dirt. Microbial air sampling devices are extensively used in Pharmaceutical and Personal care industries as well as Food, Beverage and Environmental applications.

Legal Information

MAS-100 is a registered trademark of MBV AG
MAS-100 NT is a registered trademark of MBV AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service