Key Documents
1.09126
Millipore
Shoulder bag
for use with MAS-100 Eco®
Select a Size
1 UNIT
2.100,00 kr.
2.100,00 kr.
Please contact Customer Service for Availability
Request more information
Select a Size
Change View
1 UNIT
2.100,00 kr.
About This Item
UNSPSC Code:
41122409
NACRES:
NA.74
2.100,00 kr.
Please contact Customer Service for Availability
Request more information
material
fabric bag
Agency
non-compendial
product line
MAS-100®
feature
portable
packaging
pkg of 1 unit
manufacturer/tradename
MAS-100 Eco®
application(s)
air monitoring
food and beverages
compatibility
for use with MAS-100 Eco®
General description
Shoulder bag for MAS-100 Eco® is an additional accessory. The MAS-100 Eco® is an air sampler, designed to monitor air microbiology in cleanrooms and hygienically ultra-sensitive areas. Its high recovery rate is based on the Andersen impaction principle. An integrated airflow sensor compensates for differences in pressure and temperature in the environment. The shoulder bag aids in easy and safe transport of the air sampler.
Application
Shoulder bag for MAS-100 Eco® is an additional accessory for Air Sampler that is used for microbial monitoring of ambient air in food and beverage industry. Microbial air sampling devices are extensively used in Food, Beverage as well as Environmental applications.
Legal Information
MAS-100 is a registered trademark of MBV AG
MAS-100 Eco is a registered trademark of MBV AG