Skip to Content
Merck
All Photos(1)

Documents

1.08540

Supelco

TLC sprayer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
NACRES:
NB.21

technique(s)

thin layer chromatography (TLC): suitable

storage temp.

2-30°C

General description

Our TLC accessories allow professionals to work more quickly, more efficiently, and more reliably. Thanks to our broad assortment, users will always find what they need.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service