Skip to Content
Merck
All Photos(1)

Key Documents

1.07717

Millipore

Selenite enrichment broth acc. to LEIFSON

for the selective enrichment of salmonellae

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200

form

medium granules (dehydrated (DCM))

technique(s)

microbiological culture: suitable

pH

7.0 (37 °C, 23 g/L in H2O)

solubility

23 g/L

bulk density

940 kg/m3

application(s)

clinical
microbiology

storage temp.

2-30°C

suitability

Salmonella spp.

General description

for the selective enrichment of salmonellae

Analysis Note

Appearance (clearness)clearAppearance (colour)yellowishpH-value (25 °C)6.8 - 7.2Inoculum (Escherichia coli ATCC 25922 (WDCM 00013))about 99 %Inoculum (Salmonella typhimurium ATCC 14028 (WDCM 00031))about 1 %Enrichment from a mixed inoculum after 6 hours (Escherichia coli ATCC 25922 (WDCM 00013))Enrichment from a mixed inoculum after 6 hours (Salmonella typhimurium ATCC 14028 (WDCM 00031))Enrichment from a mixed inoculum after 24 hours (Escherichia coli ATCC 25922 (WDCM 00013))≤ 10 %Enrichment from a mixed inoculum after 24 hours (Salmonella typhimurium ATCC 14028 (WDCM 00031))≥ 90 %Enrichement of Salmonella typhimurium from a mixed inoculum: Incubation: 24 Std.; 35 °C; aerobic.
Detection with Brolacin-agar (24 hrs.; 35 °C).

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service