Skip to Content
Merck
All Photos(1)

Key Documents

1.05669

Supelco

Lithium bromide

extra pure

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
3.030,00 kr.

3.030,00 kr.


Estimated to ship on28 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 KG
3.030,00 kr.

About This Item

Linear Formula:
LiBr
CAS Number:
Molecular Weight:
86.85
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21
grade:
for analytical purposes

3.030,00 kr.


Estimated to ship on28 April 2025Details


Request a Bulk Order

grade

for analytical purposes

Quality Level

pH

7 (20 °C, 10 g/L in H2O)

bp

>1265 °C/1013 hPa

mp

550 °C (lit.)

SMILES string

[Li+].[Br-]

InChI

1S/BrH.Li/h1H;/q;+1/p-1

InChI key

AMXOYNBUYSYVKV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service