Skip to Content
Merck
All Photos(1)

Documents

104648

SAFC

Cellvento® ModiFeed Gal- COMP

new
greener alternative

Chemically defined cell culture feed - EMPROVE® CCM

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213

product name

Cellvento® ModiFeed Gal- COMP, Chemically defined cell culture feed - EMPROVE® CCM

Quality Level

description

shipped with cooling packs

product line

Cellvento®

form

dry powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

cell culture | mammalian: suitable

greener alternative category

storage temp.

2-8°C

General description

Cellvento® ModiFeed Gal- COMP is part of the ModiFeed platform. It is a highly concentrated, one-part, pH neutral feed added to replenish depleted nutrients required for cellular function and to extend the production phase in fed-batch mode, while simultanously reducing galactosylation on the protein of interest. It is recommened in combination with the production media Cellvento® 4CHO and EX-CELL® Advanced CHO Fed-batch medium or other production media.
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives. This product is a Design for Sustainability (DfS) developed product. Click here for more information.

Application

Glycosylation is known to have effects on the biological activity, solubility, serum half-life, and saftey of therapeutic proteins. Control of protein glycosylation processes, such as galactosylation, while maximizing productivity is key for successful mAb process development and production. Cellvento® ModiFeed Gal- COMP was developed to increase G0(F), while maintaining or improving titer in CHO fed-batch processes.

Features and Benefits

  • Reduced complexity in fed-batch process
  • Complete feed maximizing galactosylation
  • Highly concentrated to reduce required feed volume
  • Compacted format for conventient handling
  • Higher liquid feed stability at room temperature

Quantity

Formulated to contain 143.6 grams of powder per liter of feed.

Legal Information

CELLVENTO is a registered trademark of Merck KGaA, Darmstadt, Germany
EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany
Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service