Skip to Content
Merck
All Photos(1)

Key Documents

1.04499

Supelco

ICP Multi element standard solution I certified reference material

(19 elements in diluted nitric acid), Certipur®

Sign Into View Organizational & Contract Pricing

Select a Size

1044990100
1.720,00 kr.

1.720,00 kr.


Estimated to ship on31 March 2025Details



Select a Size

Change View
1044990100
1.720,00 kr.

About This Item

UNSPSC Code:
12352200

1.720,00 kr.


Estimated to ship on31 March 2025Details


Quality Level

technique(s)

ICP: suitable

matrix

(in diluted nitric acid)

pH

0.1 (20 °C in water)

density

1.02 g/cm3 at 20 °C

application(s)

agriculture
environmental
food and beverages

format

multi-component solution

storage temp.

15-25°C

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.


Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.


Download your certificate at https://www.sigmaaldrich.com/US/en/documents-search?tab=coa  to view certified values, including uncertainty and expiry date.

Application

ICP Multi element standard solution I is a certified reference material can be used as a calibration standard in Inductively Coupled Plasma (ICP) spectroscopy

Analysis Note

Tl (thallium): 400 mg/ml
Bi (bismuth): 200 mg/l
In (indium): 200 mg/l
Pb (lead): 200 mg/l
Ga (gallium): 150 mg/ml
Al (aluminium): 100 mg/ml
Ag (silver): 50 mg/ml
Ni (nickel): 50 mg/l
Cr (chromium): 25 mg/l
Cd (cadmium): 20 mg/l
Co (cobalt): 20 mg/l
Cu (copper): 20 mg/l
Zn (zinc): 20 mg/l
B (boron): 15 mg/l
Fe (iron): 15 mg/ml
Ba (barium): 5 mg/l
Mn (manganese): 5 mg/l
Sr (strontium): 1 mg/l
Be (beryllium) : 1 mg/l

Other Notes

For a complete product listing of our Certipur® range of CRMs for ICP and AAS, technical information, and example certificates please visit our ICP & AAS standards website


For detailed information for our newly launched ICP-Multi element Standards check the flyer: Commitment to Quality: Elevating Certipur®  Standards to Certified Reference Material 


In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website

Legal Information

Certipur is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service