Skip to Content
Merck
All Photos(1)

Key Documents

1.04233

Millipore

Glycylglycine

≥99% dry basis (perchloric acid titration)

Synonym(s):

Glycylglycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8N2O3
CAS Number:
Molecular Weight:
132.12
MDL number:
UNSPSC Code:
12161703
EC Index Number:
209-127-8

Assay

≥99% dry basis (perchloric acid titration)

form

solid

storage temp.

2-30°C

InChI

1S/C4H8N2O3.ClH/c5-1-3(7)6-2-4(8)9;/h1-2,5H2,(H,6,7)(H,8,9);1H

InChI key

YHBAZQDEMYQPJL-UHFFFAOYSA-N

General description

buffer substance

Analysis Note

Assay (perchloric acid titration, calculated on dried substance): ≥ 99
Identity (IR-spectrum): passes test
UV-Absorption (250 nm, 20 g/l, 1 cm, water): ≤ 0.05
Chloride (Cl): ≤ 0.005 %
Sulfate (SO₄): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.0005 %
Na (Sodium): ≤ 0.01 %
Glycine: ≤ 0.15 %
Loss on Drying (105 °C): ≤ 1.0 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service