Skip to Content
Merck
All Photos(1)

Key Documents

1.03162

Supelco

Eschka′s mixture

GR for analysis (for determination of sulfur in coal)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352402
NACRES:
NB.21

Quality Level

form

solid

pH

12 (20 °C, 50 g/L in H2O)

solubility

80 g/L

bulk density

300 kg/m3

storage temp.

no temp limit

Analysis Note

Magnesium oxide: 65.0 - 67.0 %
Sodium carbonate: 32.0 - 34.0 %
Total sulfur (S): ≤ 0.0005 %
Loss on Drying (105 °C): ≤ 0.5 %

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service