Skip to Content
Merck
All Photos(1)

Key Documents

1.01947

Supelco

Bromine

99.9999 Suprapur®

Synonym(s):

Bromine

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
4.700,00 kr.

4.700,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
4.700,00 kr.

About This Item

CAS Number:
Molecular Weight:
159.81
MDL number:
UNSPSC Code:
41116105
EC Index Number:
231-778-1
NACRES:
NA.21
form:
liquid
Assay:
99.9999%

4.700,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

grade

for inorganic trace analysis

Quality Level

vapor pressure

233 hPa ( 20 °C)

Assay

99.9999%

form

liquid

kinematic viscosity

0.314 cSt(20 °C)

bp

58.8 °C/1013 hPa

mp

-7.3 °C

solubility

35 g/L

density

3.12 g/cm3 at 20 °C (liquid)

storage temp.

15-25°C

SMILES string

[Br-]

InChI

1S/BrH/h1H/p-1

InChI key

CPELXLSAUQHCOX-UHFFFAOYSA-M

Related Categories

General description

99.9999 Suprapur®

Application

Bromine 99.9999 Suprapur®. CAS 7726-95-6, molar mass 159.81 g/mol.

Analysis Note

Purity (metallic)≥ 99.9999 %Assay (iodometric)≥ 99.8 %Chlorine (Cl)≤ 500 ppmIodine (I)≤ 10 ppmSulfate (SO₄)≤ 5 ppmAg (Silver)≤ 0.0010 ppmAl (Aluminium)≤ 0.0050 ppmAu (Gold)≤ 0.0050 ppmBa (Barium)≤ 0.0050 ppmBe (Beryllium)≤ 0.0010 ppmBi (Bismuth)≤ 0.0050 ppmCa (Calcium)≤ 0.030 ppmCd (Cadmium)≤ 0.0010 ppmCo (Cobalt)≤ 0.0010 ppmCr (Chromium)≤ 0.0010 ppmCu (Copper)≤ 0.0010 ppmFe (Iron)≤ 0.010 ppmGa (Gallium)≤ 0.0050 ppmIn (Indium)≤ 0.0020 ppmK (Potassium)≤ 0.020 ppmLi (Lithium)≤ 0.0010 ppmMg (Magnesium)≤ 0.0050 ppmMn (Manganese)≤ 0.0010 ppmMo (Molybdenum)≤ 0.0010 ppmNa (Sodium)≤ 0.050 ppmNi (Nickel)≤ 0.0020 ppmPb (Lead)≤ 0.0020 ppmSn (Tin)≤ 0.0050 ppmSr (Strontium)≤ 0.0010 ppmTi (Titanium)≤ 0.0010 ppmTl (Thallium)≤ 0.0020 ppmV (Vanadium)≤ 0.0010 ppmZn (Zinc)≤ 0.0050 ppmZr (Zirconium)≤ 0.0010 ppmNon volatile matter≤ 10 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service