Recommended Products
Quality Level
form
granular
potency
>5000 mg/kg LD50, skin (Rabbit)
loss
≤2.0% loss on drying, 180°C
particle size
2-5 mm
pore size
2.0-2.5 nm
pH
5-8 (20 °C, 100 g/L in H2O, slurry)
bp
2230 °C
mp
>1600 °C
bulk density
750‑800 kg/m3
storage temp.
15-25°C
SMILES string
O=[Si]=O
InChI
1S/O2Si/c1-3-2
InChI key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Isolation, characterization, identification and quantification of 6-F oxyphenisatin dipropionate, a novel illegal additive, from a fruit-flavored jelly: Demonstrating the use of silica gel in chromatographic techniques for detecting illegal additives in food products, this study is vital for food safety and environmental testing laboratories (Xia et al., 2024).
Analysis Note
Loss on drying (180 °C): ≤ 2.0 %
Water absorption capacity (24 hrs.,80 % relative humidity): ≥ 25.0 %
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service