Skip to Content
Merck
All Photos(1)

Documents

1.00361

Sigma-Aldrich

ISOSLIDE Reticulin

with reference tissue for the detection of reticular fibers in histology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116124

Quality Level

IVD

for in vitro diagnostic use

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

The ISOSLIDE Reticulin Control Slides - with reference tissue for the detection of reticular fibers in histology, are intended for silver staining reticulin fibers in histology. They can be stained using the Reticulin silver plating kit according to Gordon & Sweets (Product number 1.00251) or with the laboratory′s own method. The box consists of 25 paraffin-embedded tissue specimens on glass slides, including one control slide already stained with the appropriate method and 24 unstained preparations. All tissue specimens derive from the same area of animal tissue and each box contains one batch. CE-certified, IVD product. The ISOSLIDE control slides are not quality-controlled for their application with molecular pathology methods and IHC. For more details, please see instructions for use (IFU). This product is registered as IVD and CE marked. The IFU can be downloaded from this webpage.

Application

Control slides with reference tissue for the detection of reticular fibers in histological tissue

Analysis Note

Suitability for microscopypasses testreticular fibersblack

Legal Information

ISOSLIDE is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service