Skip to Content
Merck
All Photos(1)

Key Documents

05-530-I

Sigma-Aldrich

Anti-Rad51 Antibody, clone 3C10

clone 3C10, from mouse

Synonym(s):

DNA repair protein RAD51 homolog 1, HsRAD51, hRAD51, RAD51 homolog A, Rad51

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
1.120,00 kr.
200 μG
3.090,00 kr.

1.120,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
1.120,00 kr.
200 μG
3.090,00 kr.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

1.120,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3C10, monoclonal

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), mouse (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... RAD51(5888)
mouse ... Rad51(19361)

General description

DNA repair protein RAD51 homolog 1 (UniProt Q06609; also known as BRCA1/BRCA2-containing complex subunit 5, Homolog of E. coli RecA, hRAD51, HsRAD51, RAD51 homolog A, RecA-like protein, Recombination protein A) is encoded by the RAD51 (also known as HRAD51, HsT16930, BRCC5, MRMV2, RAD51A, RECA) gene (Gene ID 5888) in human. RAD51 is a member of the RecA family recombinases. It interacts with BRCA1 and BRCA2 to activate homologous recombination. RAD51 exhibits DNA-dependent ATPase activity and participates in double-strand break (DSB) repair by unwinding DNA to form a helical nucleoprotein filament. It is highly expressed in the testis and thymus and lower expression levels are found in the small intestine, placenta, colon, pancreas, and ovaries.

Immunogen

Recombinant protein corresponding to human Rad51.

Application

Immunoprecipitation Analysis: Reported by an independent laboratory to immunoprecipitate.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
This Anti-Rad51 Antibody, clone 3C10 is validated for use in Western Blotting, Immunoprecipitation for the detection of Rad51 .

Quality

Evaluated by Western Blotting in HeLa nuclear extract.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Rad51 in 10 µg of HeLa nuclear extract.

Target description

~37 kDa observed

Linkage

Replaces: 05-530

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nataly Tarasenko et al.
Pharmaceuticals (Basel, Switzerland), 14(12) (2021-12-29)
We studied the unique inhibitor of the histone deacetylases (HDAC) valproate-valpromide of acyclovir (AN446) that upon metabolic degradation release the HDAC inhibitor (HDACI) valproic acid (VPA). Among the HDAC inhibitors that we have tested, only AN446, and to a lesser
Michelle Mac et al.
Journal of virology, 97(5), e0020123-e0020123 (2023-05-08)
The human papillomavirus (HPV) life cycle takes place in the stratified epithelium, with the productive phase being activated by epithelial differentiation. The HPV genome is histone-associated, and the life cycle is epigenetically regulated, in part, by histone tail modifications that
Kavi Mehta et al.
mBio, 9(1) (2018-02-15)
High-risk human papillomaviruses (HPVs) activate the ataxia telangiectasia mutated-dependent (ATM) DNA damage response as well as the ataxia telangiectasia mutated-dependent DNA-related (ATR) pathway in the absence of external DNA damaging agents for differentiation-dependent genome amplification. Through the use of comet
Jessica D Tischler et al.
Nature communications, 15(1), 866-866 (2024-01-30)
Homologous recombination (HR) plays critical roles in repairing lesions that arise during DNA replication and is thus essential for viability. RAD51 plays important roles during replication and HR, however, how RAD51 is regulated downstream of nucleofilament formation and how the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service