03-250
RIPAb+ Ago3 Antibody
clone 4B1-F6, from mouse
Synonym(s):
eIF-2C 3, eIF2C 3, FLJ12765, MGC86946
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified antibody
clone
4B1-F6, monoclonal
species reactivity
human
manufacturer/tradename
RIPAb+
Upstate®
technique(s)
RIP: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
isotype
IgG2aκ
NCBI accession no.
UniProt accession no.
shipped in
dry ice
Related Categories
General description
RIPAb+ antibodies are evaluated using the RNA Binding Protein Immunoprecipitation (RIP) assay. Each RIPAb+ antibody set includes a negative control antibody to ensure specificity of the RIP reaction and is verified for the co-immunoprecipitation of RNA associated specifically with the immunoprecipitated RNA-binding protein of interest. Where appropriate, the RIPAb+ set also includes quantitative RT-PCR control primers (RIP Primers) to biologically validate your IP results by successfully co-precipitating the specific RNA targets, such as messenger RNAs. The qPCR protocol and primer sequences are provided, allowing researchers to validate RIP protocols when using the antibody in their experimental context. If a target specific assay is not provided, the RIPAb+ kit is validated using an automated microfluidics-based assay by enrichment of detectable RNA over control immunoprecipitation.
Ago3 (Eukaryotic translation initiation factor 2C 3) belongs to the argonaute family and contains a PAZ and a Piwi domain. Ago3 plays a role in the RNA interference (RNAi) pathway, which is important for heterochromatin formation and accurate chromosome segregation.
Ago3 (Eukaryotic translation initiation factor 2C 3) belongs to the argonaute family and contains a PAZ and a Piwi domain. Ago3 plays a role in the RNA interference (RNAi) pathway, which is important for heterochromatin formation and accurate chromosome segregation.
Specificity
This antibody has been shown to not cross react with Ago1, Ago2, or Ago4 (Azuma-Mukai A et al. (2008) Proc. Natl. Acad. Sci. USA 105: 7964-7969).
Immunogen
Recombinant protein corresponding to human Ago3
Application
Ago3 (Eukaryotic translation initiation factor 2C 3) belongs to the argonaute family & contains a PAZ & a Piwi domain. Ago3 plays a role in the RNA interference (RNAi) pathway.
RNA Binding Protein Immunoprecipitation (microRNA):
Representative lot data.
RIP Lysate prepared from 293 cells (2 X 107 cell equivalents per IP) were subjected to immunoprecipitation using 5 µg of either a normal mouse IgG (Cat. # CS200621), or Anti-Ago 3 antibody (Cat. # CS207366) and the Magna RIP RNA-Binding Protein Immunoprecipitation Kit (Cat. # 17-700).
Successful immunoprecipitation of Ago 3-associated microRNA was verified by TaqMan microRNA assay, hsa-miR-15a. (Figure2). Please refer to the Magna RIP (Cat. # 17-700) or EZ-Magna RIP (Cat. # 17-701) protocol for experimental details.
Immunocytochemistry Analysis:
Representative lot data.
Confocal fluorescent analysis of HeLa cells using Anti-Ago3, clone 4B1-F6 (Red). Actin filaments have been labeled with Alexa Fluor 488 dye-Phalloidin (Green). Nucleus is stained with DAPI (Blue). This antibody positively stains the cytoplasm.
(Alexa Fluor is a registered trademark of Life Technologies.)
Immunoprecipitation Analysis: A representative lot was used by an independent laboratory in HeLa cells. (Azuma-Mukai, A., et al. (2008). Proc Natl Acad Sci USA. 105(23):7964-7969.)
Representative lot data.
RIP Lysate prepared from 293 cells (2 X 107 cell equivalents per IP) were subjected to immunoprecipitation using 5 µg of either a normal mouse IgG (Cat. # CS200621), or Anti-Ago 3 antibody (Cat. # CS207366) and the Magna RIP RNA-Binding Protein Immunoprecipitation Kit (Cat. # 17-700).
Successful immunoprecipitation of Ago 3-associated microRNA was verified by TaqMan microRNA assay, hsa-miR-15a. (Figure2). Please refer to the Magna RIP (Cat. # 17-700) or EZ-Magna RIP (Cat. # 17-701) protocol for experimental details.
Immunocytochemistry Analysis:
Representative lot data.
Confocal fluorescent analysis of HeLa cells using Anti-Ago3, clone 4B1-F6 (Red). Actin filaments have been labeled with Alexa Fluor 488 dye-Phalloidin (Green). Nucleus is stained with DAPI (Blue). This antibody positively stains the cytoplasm.
(Alexa Fluor is a registered trademark of Life Technologies.)
Immunoprecipitation Analysis: A representative lot was used by an independent laboratory in HeLa cells. (Azuma-Mukai, A., et al. (2008). Proc Natl Acad Sci USA. 105(23):7964-7969.)
Research Category
Epigenetics & Nuclear Function
Epigenetics & Nuclear Function
Research Sub Category
RNA Binding Protein (RBP)
RNA Binding Protein (RBP)
Packaging
10 assays per set. Recommended use: ~5 µg of antibody per RIP (dependent upon biological context).
Quality
RNA Binding Protein Immunoprecipitation:
Representative lot data
RIP Lysate prepared from 293 cells (2 X 10E7 cell equivalents per IP) were subjected to immunoprecipitation using 5 µg of either a normal mouse IgG , or 5 µg Anti-Ago 3 antibody and the Magna RIP® RNA-Binding Protein Immunoprecipitation Kit (Cat. # 17-700).
Successful immunoprecipitation of Argonaute 3-associated RNA was verified by qPCR using RIP Primers IGF2, (Figure 1).
Please refer to the Magna RIP (Cat. # 17-700) or EZ-Magna RIP (Cat. # 17-701) protocol for experimental details.
Representative lot data
RIP Lysate prepared from 293 cells (2 X 10E7 cell equivalents per IP) were subjected to immunoprecipitation using 5 µg of either a normal mouse IgG , or 5 µg Anti-Ago 3 antibody and the Magna RIP® RNA-Binding Protein Immunoprecipitation Kit (Cat. # 17-700).
Successful immunoprecipitation of Argonaute 3-associated RNA was verified by qPCR using RIP Primers IGF2, (Figure 1).
Please refer to the Magna RIP (Cat. # 17-700) or EZ-Magna RIP (Cat. # 17-701) protocol for experimental details.
Target description
~96 kDa
Physical form
Anti-Ago3 (Mouse Monoclonal). One vial containing 50 µg of protein G purified monoclonal in buffer containing 0.1 M Tris-glycine (pH 7.4), 150mM NaCl, 0.05% sodium azide before the addition of glycerol to 30%. Store at -20° C.
Concentration: 0.7 mg/mL
Normal mouse IgG. One vial containing 125 µg purified mouse IgG in 125 µL storage buffer containing 0.1% sodium azide. Store at -20°C.
RIP Primer IGF2. One vial containing 75 μL of 5 μM of each primer specific for human IGF2 mRNA. Store at
-20°C.
FOR: GCG GCT TCT ACT TCA GCA G
REV: CAG GTG TCA TAT TGG AAG AAC
Concentration: 0.7 mg/mL
Normal mouse IgG. One vial containing 125 µg purified mouse IgG in 125 µL storage buffer containing 0.1% sodium azide. Store at -20°C.
RIP Primer IGF2. One vial containing 75 μL of 5 μM of each primer specific for human IGF2 mRNA. Store at
-20°C.
FOR: GCG GCT TCT ACT TCA GCA G
REV: CAG GTG TCA TAT TGG AAG AAC
Format: Purified
Protein G Purified
Storage and Stability
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.
Analysis Note
Control
Includes normal rabbit IgG and primers specific for human IGF2 mRNA.
Includes normal rabbit IgG and primers specific for human IGF2 mRNA.
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Legal Information
MAGNA RIP is a registered trademark of Merck KGaA, Darmstadt, Germany
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of cellular biochemistry, 116(12), 2744-2754 (2015-07-25)
Non-coding RNAs (ncRNAs) play key roles in epigenetic events. However, the exact mechanism of ncRNA guidance, particularly piwi-interacting RNAs (piRNAs), for the targeting of epigenetic regulatory factors to specific gene regions is unclear. Although piRNA function was first established in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service