Skip to Content
Merck
All Photos(1)

Key Documents

03-0134-00

Millipore

SMC® Mouse IL-10 Discovery Immunoassay Kit

Synonym(s):

mouse IL 10 immunoassay kit, mouse IL-10 Single Molecule Counting Kit, mouse IL-10 ultra-sensitive immunoassay, mouse Interleukin 10 immunoassay kit

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
7.510,00 kr.

7.510,00 kr.


Estimated to ship on31 March 2025Details


Request more information

Select a Size

Change View
96 WELLS
7.510,00 kr.

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.47

7.510,00 kr.


Estimated to ship on31 March 2025Details


Request more information

species reactivity

mouse

Quality Level

manufacturer/tradename

SMC®

assay range

accuracy: 80-120%
(minimum of 10 samples spiked with acceptable recovery)

technique(s)

ELISA: suitable
single molecule counting: suitable

General description

Interleukin-10 (IL-10) is a cytokine with both stimulatory and suppressive effects on the immune system. The protein originates from several cell lines, including Th2 cells, mast cells, macrophages, monocytes and B-cells. Its main functions are to impede inflammatory responses and to inhibit synthesis of other pro-inflammatory cytokines, such as IL-2, IL-3, GM-CSF, IFN-γ and TNF-α. Besides its anti-inflammatory capabilities, IL-10 also regulates the production of natural killer cells and lymphocytes. Due to its multiple roles, proper function of IL-10 is required to maintain a healthy immune system. Improper function of IL-10 can result in disease development, as over expression of IL-10 has been linked to lymphoma, whereas IL-10 deficiency has implications for Crohn′s disease.
The SMC® Mouse IL-10 Discovery Immunoassay Kit contains all reagents required to perform the assay. This assay takes advantage of a standard immunoassay workflow configured in a standard plate format. Assay protocols are similar to existing sandwich ELISA methods with two key differences: 1) Elution buffer disrupts the sandwich, separating the labeled detection antibody for quantification. 2) The SMC® immunoassay system detects analytes using Single Molecule Counting (SMC®) technology.

Application

Research Category
Inflammation & Immunology
Research Sub Category
Cytokines & Cytokine Receptors
The sensitivity of Single Molecule Counting (SMC®) technology gives the SMC® Mouse IL-10 Discovery Immunoassay has unparalleled performance in quantifying IL-10 in mouse serum.
Used to detect/quantify:IL-10

Components

Assay Buffer
Mouse IL-10 Coating Antibody
Assay Plate
10X PBS
Blocking Buffer
Standard Diluent
Mouse IL-10 Detection Antibody
Mouse IL-10 Standard
SMC® Mouse IL-10 Immunoassay Kit Instructions
10X Wash Buffer
Buffer C
Elution Buffer B

Analysis Note

Low-end CV% Range: 0-11% Low-end CV% Average: 6%

Legal Information

SMC is a registered trademark of Singulex

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service