Skip to Content
Merck
All Photos(1)

Key Documents

MAT490083004

Matrix Scientific

Methyl 4,4,4-trifluorocrotonate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
154.09
MDL number:

bp

95

density

1.237

Iupac Name

methyl (2E)-4,4,4-trifluoro-2-butenoate

Inchi Code

1S/C5H5F3O2/c1-10-4(9)2-3-5(6,7)8/h2-3H,1H3

InChI key

DMMZYYLXAGRBDO-UHFFFAOYSA-N

Purity

95%

Country of Origin

US

Product Link

https://www.matrixscientific.com/002412.html

Flash Point

32C

Boiling point

95

density

1.237

MSDS

https://www.matrixscientific.com/002412.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library