Skip to Content
Merck
All Photos(1)

Documents

MAN182108931

Manchester Organics

4-Benzylbenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
196.25
MDL number:

Iupac Name

4-benzylbenzaldehyde

Inchi Code

1S/C14H12O/c15-11-14-8-6-13(7-9-14)10-12-4-2-1-3-5-12/h1-9,11H,10H2

InChI key

DHARILAASAAGJJ-UHFFFAOYSA-N

Purity

85%

Country of Origin

GB

Product Link

https://www.manchesterorganics.com/G18314

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service