LIFH97ED0190
Life Chemicals Inc.
2-(pyridin-4-yloxy)aniline
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
2-(4-pyridinyloxy)aniline
Inchi Code
1S/C11H10N2O/c12-10-3-1-2-4-11(10)14-9-5-7-13-8-6-9/h1-8H,12H2
InChI key
NIRPDVZBDYXCSS-UHFFFAOYSA-N
Purity
95%
Country of Origin
UA
Physical Form
solid
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.