Skip to Content
Merck
All Photos(1)

Key Documents

KEY465197071

Key Organics / BIONET

3-{2-[3-(trifluoromethyl)phenyl]hydrazin-1-ylidene}pentane-2,4-dione

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
272.23
MDL number:

Iupac Name

2,3,4-pentanetrione 3-{[3-(trifluoromethyl)phenyl]hydrazone}

Inchi Code

1S/C12H11F3N2O2/c1-7(18)11(8(2)19)17-16-10-5-3-4-9(6-10)12(13,14)15/h3-6,16H,1-2H3

InChI key

QXNTVEFIHAGSDS-UHFFFAOYSA-N

Purity

90%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/5M-922

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

MSDS

https://www.keyorganics.net/sds/5M-922


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library