Skip to Content
Merck
All Photos(1)

Key Documents

KEY001132542

Key Organics / BIONET

ethyl 3-benzamido-2-oxo-6-phenyl-2H-pyran-5-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
363.37
MDL number:

Pricing and availability is not currently available.

mp

147 - 149

Iupac Name

ethyl 3-(benzoylamino)-2-oxo-6-phenyl-2H-pyran-5-carboxylate

Inchi Code

1S/C21H17NO5/c1-2-26-20(24)16-13-17(22-19(23)15-11-7-4-8-12-15)21(25)27-18(16)14-9-5-3-6-10-14/h3-13H,2H2,1H3,(H,22,23)

InChI key

JWSNPYGLRSSRSX-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/2E-917

Melting Point

147 - 149

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library