ENAH961CE872
Enamine
4-(3,4-dichlorophenyl)oxane-2,6-dione
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
mp
132-135
Iupac Name
4-(3,4-dichlorophenyl)dihydro-2H-pyran-2,6(3H)-dione
Inchi Code
1S/C11H8Cl2O3/c12-8-2-1-6(3-9(8)13)7-4-10(14)16-11(15)5-7/h1-3,7H,4-5H2
InChI key
UOLHGGKIAWBSPT-UHFFFAOYSA-N
Purity
95
Country of Origin
UA
Product Link
https://www.enaminestore.com/catalog/EN300-366279
Melting Point
132-135
MSDS
https://www.enaminestore.com/catalog/EN300-366279
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.