Skip to Content
Merck
All Photos(1)

Key Documents

ENAH315519EA

Enamine

4-(trifluoromethyl)-1,3-thiazole-2-sulfonyl fluoride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
235.18
MDL number:

Iupac Name

4-(trifluoromethyl)thiazole-2-sulfonyl fluoride

Inchi Code

1S/C4HF4NO2S2/c5-4(6,7)2-1-12-3(9-2)13(8,10)11/h1H

InChI key

OUXWSGCAVOBFKD-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-6482938

MSDS

https://www.enaminestore.com/catalog/EN300-6482938


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library