ENAH03A179D8
Enamine
5-chloro-6-fluoropyridine-3-sulfonyl fluoride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
5-chloro-6-fluoropyridine-3-sulfonyl fluoride
Inchi Code
1S/C5H2ClF2NO2S/c6-4-1-3(12(8,10)11)2-9-5(4)7/h1-2H
InChI key
PARZZSYIHCEZCS-UHFFFAOYSA-N
Purity
95
Country of Origin
UA
Product Link
https://www.enaminestore.com/catalog/EN300-1167006
MSDS
https://www.enaminestore.com/catalog/EN300-1167006
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.